Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NAT9 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NAT9 - N-acetyltransferase 9 (putative) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM104 - transmembrane protein 104 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321264.1 | Intron | NP_001308193.1 | ||||
NM_017728.3 | Intron | NP_060198.3 | ||||
XM_006721963.1 | Intron | XP_006722026.1 | ||||
XM_017024797.1 | Intron | XP_016880286.1 | ||||
XM_017024798.1 | Intron | XP_016880287.1 | ||||
XM_017024799.1 | Intron | XP_016880288.1 |