Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612185 MIM: 608755 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CASKIN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CASKIN2 - CASK interacting protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LLGL2 - LLGL2, scribble cell polarity complex component | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TSEN54 - tRNA splicing endonuclease subunit 54 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207346.2 | 146 | Missense Mutation | CAG,CAT | Q,H 38 | NP_997229.2 | |
XM_005257229.3 | 146 | Missense Mutation | CAG,CAT | Q,H 38 | XP_005257286.1 |
Set Membership: |
HapMap |