Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATACAATGAAGCCTAATCATCCTGA[A/G]TATAGGGCAGGGCAATTAAAGTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC5A10 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC5A10 - solute carrier family 5 member 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042450.2 | Intron | NP_001035915.1 | ||||
NM_001270648.1 | Intron | NP_001257577.1 | ||||
NM_001270649.1 | Intron | NP_001257578.1 | ||||
NM_001282417.1 | Intron | NP_001269346.1 | ||||
NM_152351.4 | Intron | NP_689564.3 | ||||
XM_011523652.1 | Intron | XP_011521954.1 | ||||
XM_011523653.2 | Intron | XP_011521955.1 | ||||
XM_017024191.1 | Intron | XP_016879680.1 |
Set Membership: |
HapMap |