Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601522 MIM: 611802 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRB7 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GRB7 - growth factor receptor bound protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001030002.2 | 261 | Intron | NP_001025173.1 | |||
NM_001242442.1 | 261 | Intron | NP_001229371.1 | |||
NM_001242443.1 | 261 | Intron | NP_001229372.1 | |||
NM_005310.3 | 261 | Intron | NP_005301.2 | |||
XM_017024536.1 | 261 | UTR 5 | XP_016880025.1 | |||
XM_017024537.1 | 261 | Intron | XP_016880026.1 | |||
XM_017024538.1 | 261 | Intron | XP_016880027.1 |
MIEN1 - migration and invasion enhancer 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |