Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGCGATGGCACCCACAGGAGTGA[G/T]CGCGTGCAGAAGGCAGAAGTGGGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611933 MIM: 600328 MIM: 600346 | ||||||||||||||||||||
Literature Links: |
CISD3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CISD3 - CDGSH iron sulfur domain 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136498.1 | 450 | Missense Mutation | GAG,GAT | E,D 116 | NP_001129970.1 |
MLLT6 - MLLT6, PHD finger domain containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PCGF2 - polycomb group ring finger 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007144.2 | 450 | Intron | NP_009075.1 | |||
XM_005257640.2 | 450 | Intron | XP_005257697.1 | |||
XM_005257641.4 | 450 | Intron | XP_005257698.1 | |||
XM_005257642.3 | 450 | Intron | XP_005257699.1 | |||
XM_017025016.1 | 450 | Intron | XP_016880505.1 |