Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601440 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C17orf67 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C17orf67 - chromosome 17 open reading frame 67 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DGKE - diacylglycerol kinase epsilon | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003647.2 | Intron | NP_003638.1 | ||||
XM_011525394.2 | Intron | XP_011523696.1 | ||||
XM_011525395.2 | Intron | XP_011523697.1 | ||||
XM_011525396.2 | Intron | XP_011523698.1 | ||||
XM_017025243.1 | Intron | XP_016880732.1 | ||||
XM_017025244.1 | Intron | XP_016880733.1 | ||||
XM_017025245.1 | Intron | XP_016880734.1 |