Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607833 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FLJ45513 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FLJ45513 - uncharacterized LOC729220 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242791.1 | 2052 | UTR 3 | NP_001229720.1 |
TAC4 - tachykinin 4 (hemokinin) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077503.1 | 2052 | Intron | NP_001070971.1 | |||
NM_001077504.1 | 2052 | Intron | NP_001070972.1 | |||
NM_001077505.1 | 2052 | Intron | NP_001070973.1 | |||
NM_001077506.1 | 2052 | Intron | NP_001070974.1 | |||
NM_170685.2 | 2052 | Intron | NP_733786.2 |