Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGACGGTGCCCCATAGATGGCCA[A/G]CTTTCCCCCGAGGGTCAACGAGAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610091 | ||||||||||||||||||||
Literature Links: |
MIR4522 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR4522 - microRNA 4522 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WSB1 - WD repeat and SOCS box containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015626.8 | 324 | Missense Mutation | AAC,AGC | N,S 3 | NP_056441.6 | |
NM_134265.2 | 324 | Missense Mutation | AAC,AGC | N,S 3 | NP_599027.1 | |
XM_005257963.3 | 324 | UTR 5 | XP_005258020.1 | |||
XM_017024433.1 | 324 | Missense Mutation | AAC,AGC | N,S 3 | XP_016879922.1 | |
XM_017024434.1 | 324 | UTR 5 | XP_016879923.1 | |||
XM_017024435.1 | 324 | UTR 5 | XP_016879924.1 | |||
XM_017024436.1 | 324 | UTR 5 | XP_016879925.1 |