Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGAGGTTACTAACCAGCTATCAG[C/T]ATATTATTTTCGATTTGTGTCTAGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600940 MIM: 609735 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LIG3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LIG3 - DNA ligase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002311.4 | 3506 | Intron | NP_002302.2 | |||
NM_013975.3 | 3506 | Intron | NP_039269.2 | |||
XM_005257970.3 | 3506 | Intron | XP_005258027.1 | |||
XM_006721896.3 | 3506 | Intron | XP_006721959.1 | |||
XM_017024624.1 | 3506 | Intron | XP_016880113.1 |
RAD51L3-RFFL - RAD51L3-RFFL readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RFFL - ring finger and FYVE-like domain containing E3 ubiquitin protein ligase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017368.1 | 3506 | UTR 3 | NP_001017368.1 |
Set Membership: |
HapMap |