Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CCDC178 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC178 - coiled-coil domain containing 178 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105528.1 | Intron | NP_001098998.1 | ||||
NM_001308126.1 | Intron | NP_001295055.1 | ||||
NM_198995.2 | Intron | NP_945346.2 | ||||
XM_011525948.1 | Intron | XP_011524250.1 | ||||
XM_011525951.1 | Intron | XP_011524253.1 | ||||
XM_011525954.2 | Intron | XP_011524256.1 | ||||
XM_011525955.1 | Intron | XP_011524257.1 | ||||
XM_017025721.1 | Intron | XP_016881210.1 | ||||
XM_017025722.1 | Intron | XP_016881211.1 | ||||
XM_017025723.1 | Intron | XP_016881212.1 | ||||
XM_017025724.1 | Intron | XP_016881213.1 | ||||
XM_017025725.1 | Intron | XP_016881214.1 |