Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAAAATCCTGGTCTGTGGAAGGG[C/T]CACAATCACAGTGACCCTGTGAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176390 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PSG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PSG1 - pregnancy specific beta-1-glycoprotein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184825.1 | Intron | NP_001171754.1 | ||||
NM_001184826.1 | Intron | NP_001171755.1 | ||||
NM_001297773.1 | Intron | NP_001284702.1 | ||||
NM_006905.2 | Intron | NP_008836.2 | ||||
XM_005259065.3 | Intron | XP_005259122.1 | ||||
XM_011527125.1 | Intron | XP_011525427.1 | ||||
XM_017026994.1 | Intron | XP_016882483.1 |