Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176398 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PSG9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PSG9 - pregnancy specific beta-1-glycoprotein 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301707.1 | Intron | NP_001288636.1 | ||||
NM_001301708.1 | Intron | NP_001288637.1 | ||||
NM_001301709.1 | Intron | NP_001288638.1 | ||||
NM_002784.4 | Intron | NP_002775.3 | ||||
XM_005259075.3 | Intron | XP_005259132.1 | ||||
XM_017027004.1 | Intron | XP_016882493.1 | ||||
XM_017027005.1 | Intron | XP_016882494.1 | ||||
XM_017027006.1 | Intron | XP_016882495.1 | ||||
XM_017027007.1 | Intron | XP_016882496.1 |