Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGCAGGTAGACTTCAGCCGCTAT[A/G]ACTTCTTCCTGCTGGTCTCCCCCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613176 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IRGC PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IRGC - immunity related GTPase cinema | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019612.3 | 584 | Missense Mutation | AAC,GAC | N,D 135 | NP_062558.1 | |
XM_005259058.3 | 584 | Missense Mutation | AAC,GAC | N,D 172 | XP_005259115.1 | |
XM_006723283.3 | 584 | Missense Mutation | AAC,GAC | N,D 135 | XP_006723346.1 | |
XM_011527118.2 | 584 | Missense Mutation | AAC,GAC | N,D 135 | XP_011525420.1 |
SMG9 - SMG9, nonsense mediated mRNA decay factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |