Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
GGGCCACCCAGTGCCGCCGGCGCAG[C/T]GGCAGTGACAGCAGCCCCAGCGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614545 MIM: 615324 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASPDH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ASPDH - aspartate dehydrogenase domain containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EMC10 - ER membrane protein complex subunit 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175063.5 | 459 | Intron | NP_778233.4 | |||
NM_206538.3 | 459 | Intron | NP_996261.1 | |||
XM_006723163.3 | 459 | Intron | XP_006723226.1 | |||
XM_011526818.2 | 459 | Intron | XP_011525120.1 |
JOSD2 - Josephin domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270639.1 | 459 | Silent Mutation | CCA,CCG | P,P 120 | NP_001257568.1 | |
NM_001270640.1 | 459 | Silent Mutation | CCA,CCG | P,P 120 | NP_001257569.1 | |
NM_001270641.1 | 459 | Silent Mutation | CCA,CCG | P,P 78 | NP_001257570.1 | |
NM_001270686.1 | 459 | Silent Mutation | CCA,CCG | P,P 120 | NP_001257615.1 | |
NM_138334.3 | 459 | Silent Mutation | CCA,CCG | P,P 120 | NP_612207.1 | |
XM_011526434.2 | 459 | Silent Mutation | CCA,CCG | P,P 120 | XP_011524736.1 |