Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGTCTCTCTCCTGTATGAGTCCT[C/T]TGATGGACAATGAGCTTTTGCTTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605422 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF350 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF350 - zinc finger protein 350 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021632.3 | 1566 | Silent Mutation | NP_067645.3 | |||
XM_011527196.1 | 1566 | Silent Mutation | XP_011525498.1 | |||
XM_017027094.1 | 1566 | Missense Mutation | XP_016882583.1 | |||
XM_017027095.1 | 1566 | Silent Mutation | XP_016882584.1 | |||
XM_017027096.1 | 1566 | Silent Mutation | XP_016882585.1 | |||
XM_017027097.1 | 1566 | Silent Mutation | XP_016882586.1 | |||
XM_017027098.1 | 1566 | Silent Mutation | XP_016882587.1 | |||
XM_017027099.1 | 1566 | Silent Mutation | XP_016882588.1 | |||
XM_017027100.1 | 1566 | Silent Mutation | XP_016882589.1 | |||
XM_017027101.1 | 1566 | Silent Mutation | XP_016882590.1 | |||
XM_017027102.1 | 1566 | Silent Mutation | XP_016882591.1 | |||
XM_017027103.1 | 1566 | Silent Mutation | XP_016882592.1 | |||
XM_017027104.1 | 1566 | Silent Mutation | XP_016882593.1 |
ZNF350-AS1 - ZNF350 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |