Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGAGGGGCTGCTCCCTCACAGAG[G/T]GAGCCTCTAAGCCCACCAGGCGGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605546 | ||||||||||||||||||||
Literature Links: |
GP6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GP6 - glycoprotein VI platelet | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RDH13 - retinol dehydrogenase 13 (all-trans/9-cis) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145971.1 | 1150 | Missense Mutation | CAC,CCC | H,P 322 | NP_001139443.1 | |
NM_138412.3 | 1150 | Missense Mutation | CAC,CCC | H,P 251 | NP_612421.1 | |
XM_005258473.1 | 1150 | Missense Mutation | CAC,CCC | H,P 322 | XP_005258530.1 | |
XM_011526407.1 | 1150 | Missense Mutation | CAC,CCC | H,P 322 | XP_011524709.1 | |
XM_011526408.2 | 1150 | Intron | XP_011524710.1 |