Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGGCGGTATGCCGCCAAACGCTT[C/T]CGCAAAGCTCAGTGTCCCATTGTGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603630 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR4754 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MIR4754 - microRNA 4754 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF225 - ring finger protein 225 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPS5 - ribosomal protein S5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009.3 | Intron | NP_001000.2 |
Set Membership: |
HapMap |