Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGGAGCCCCGCCTGCTGCCCTGT[C/T]TGCACTCGGCCTGTAGTGCCTGCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610893 MIM: 601742 | ||||||||||||||||||||
Literature Links: |
CHMP2A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHMP2A - charged multivesicular body protein 2A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6807 - microRNA 6807 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRIM28 - tripartite motif containing 28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005762.2 | 539 | Silent Mutation | CTG,TTG | L,L 84 | NP_005753.1 |
ZBTB45 - zinc finger and BTB domain containing 45 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |