Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603972 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF43 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF43 - zinc finger protein 43 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256648.1 | Intron | NP_001243577.1 | ||||
NM_001256649.1 | Intron | NP_001243578.1 | ||||
NM_001256650.1 | Intron | NP_001243579.1 | ||||
NM_001256651.1 | Intron | NP_001243580.1 | ||||
NM_001256653.1 | Intron | NP_001243582.1 | ||||
NM_001256654.1 | Intron | NP_001243583.1 | ||||
NM_003423.3 | Intron | NP_003414.2 | ||||
XM_011528257.2 | Intron | XP_011526559.1 | ||||
XM_011528259.2 | Intron | XP_011526561.1 | ||||
XM_017027207.1 | Intron | XP_016882696.1 | ||||
XM_017027208.1 | Intron | XP_016882697.1 | ||||
XM_017027209.1 | Intron | XP_016882698.1 | ||||
XM_017027210.1 | Intron | XP_016882699.1 | ||||
XM_017027211.1 | Intron | XP_016882700.1 | ||||
XM_017027212.1 | Intron | XP_016882701.1 | ||||
XM_017027213.1 | Intron | XP_016882702.1 | ||||
XM_017027214.1 | Intron | XP_016882703.1 | ||||
XM_017027215.1 | Intron | XP_016882704.1 | ||||
XM_017027216.1 | Intron | XP_016882705.1 |