Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607963 MIM: 606154 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MBD3L1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MBD3L1 - methyl-CpG binding domain protein 3 like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MUC16 - mucin 16, cell surface associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024690.2 | Intron | NP_078966.2 | ||||
XM_017027486.1 | Intron | XP_016882975.1 | ||||
XM_017027487.1 | Intron | XP_016882976.1 | ||||
XM_017027488.1 | Intron | XP_016882977.1 | ||||
XM_017027489.1 | Intron | XP_016882978.1 | ||||
XM_017027490.1 | Intron | XP_016882979.1 | ||||
XM_017027491.1 | Intron | XP_016882980.1 | ||||
XM_017027492.1 | Intron | XP_016882981.1 | ||||
XM_017027493.1 | Intron | XP_016882982.1 | ||||
XM_017027494.1 | Intron | XP_016882983.1 | ||||
XM_017027495.1 | Intron | XP_016882984.1 | ||||
XM_017027496.1 | Intron | XP_016882985.1 | ||||
XM_017027497.1 | Intron | XP_016882986.1 | ||||
XM_017027498.1 | Intron | XP_016882987.1 | ||||
XM_017027499.1 | Intron | XP_016882988.1 | ||||
XM_017027500.1 | Intron | XP_016882989.1 | ||||
XM_017027501.1 | Intron | XP_016882990.1 |
Set Membership: |
HapMap |