Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCAGAGCAGCTGGCACGTGACTTC[C/T]GCATCCCTGACAAGAGGTAGGTGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 176884 MIM: 608550 | ||||||||||||||||||||
Literature Links: |
PTPRA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PTPRA - protein tyrosine phosphatase, receptor type A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002836.3 | 531 | UTR 5 | NP_002827.1 | |||
NM_080840.2 | 531 | Intron | NP_543030.1 | |||
NM_080841.2 | 531 | Intron | NP_543031.1 |
VPS16 - VPS16, CORVET/HOPS core subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022575.3 | 531 | Missense Mutation | CGC,TGC | R,C 720 | NP_072097.2 | |
NM_080413.2 | 531 | Missense Mutation | CGC,TGC | R,C 576 | NP_536338.1 |