Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGAACAGCCTGGTACAATGAGAT[A/G]TTTTCTTAAGAAAGAACTTCCCAGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601988 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LIMK2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LIMK2 - LIM domain kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031801.1 | Intron | NP_001026971.1 | ||||
NM_005569.3 | Intron | NP_005560.1 | ||||
NM_016733.2 | Intron | NP_057952.1 |