Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602721 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DMC1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DMC1 - DNA meiotic recombinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278208.1 | Intron | NP_001265137.1 | ||||
NM_007068.3 | Intron | NP_008999.2 | ||||
XM_006724112.2 | Intron | XP_006724175.1 | ||||
XM_011529834.1 | Intron | XP_011528136.1 | ||||
XM_011529835.2 | Intron | XP_011528137.1 | ||||
XM_011529837.1 | Intron | XP_011528139.1 | ||||
XM_011529838.1 | Intron | XP_011528140.1 | ||||
XM_017028542.1 | Intron | XP_016884031.1 |
Set Membership: |
HapMap |