Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGTCAGTATTACTTTCCTCCTGG[C/T]GTCATAAGTTACAATGGATATTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300206 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL1RAPL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IL1RAPL1 - interleukin 1 receptor accessory protein like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014271.3 | Intron | NP_055086.1 | ||||
XM_017029240.1 | Intron | XP_016884729.1 | ||||
XM_017029241.1 | Intron | XP_016884730.1 |
Set Membership: |
HapMap |