Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCCTTTCAAGAAAGGGCTGTGGT[A/C]TGTGGAAGGTGTCAGGAACATATTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 426000 | ||||||||||||||||||||
Literature Links: |
KDM5D PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KDM5D - lysine demethylase 5D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146705.1 | 4960 | Missense Mutation | GAC,TAC | D,Y 1530 | NP_001140177.1 | |
NM_001146706.1 | 4960 | Missense Mutation | GAC,TAC | D,Y 1442 | NP_001140178.1 | |
NM_004653.4 | 4960 | Missense Mutation | GAC,TAC | D,Y 1499 | NP_004644.2 | |
XM_005262560.2 | 4960 | Missense Mutation | GAC,TAC | D,Y 1454 | XP_005262617.1 | |
XM_005262561.2 | 4960 | Missense Mutation | GAC,TAC | D,Y 1422 | XP_005262618.1 | |
XM_011531468.2 | 4960 | Missense Mutation | GAC,TAC | D,Y 1473 | XP_011529770.1 |