Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609249 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HERC6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HERC6 - HECT and RLD domain containing E3 ubiquitin protein ligase family member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001165136.1 | Intron | NP_001158608.1 | ||||
NM_017912.3 | Intron | NP_060382.3 | ||||
XM_005263083.3 | Intron | XP_005263140.1 | ||||
XM_011532053.2 | Intron | XP_011530355.1 | ||||
XM_017008333.1 | Intron | XP_016863822.1 |
Set Membership: |
HapMap |