Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCAGTGGAATAAGAACAGTCAAC[A/G]TAGGAAGAGACAGAAACATTCTTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610662 | ||||||||||||||||||||
Literature Links: |
LOC101929134 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC101929134 - uncharacterized LOC101929134 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIGY - phosphatidylinositol glycan anchor biosynthesis class Y | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042616.2 | 564 | Missense Mutation | ACG,ATG | T,M 7 | NP_001036081.1 |
PYURF - PIGY upstream reading frame | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032906.4 | 564 | UTR 3 | NP_116295.1 |