Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCACCGTGATCCTGTTAGATATC[G/T]AAGGTACCACAACCCCGATTGCTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607137 | ||||||||||||||||||||
Literature Links: |
ENOPH1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ENOPH1 - enolase-phosphatase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001292017.1 | 402 | UTR 5 | NP_001278946.1 | |||
NM_021204.4 | 402 | Nonsense Mutation | GAA,TAA | E,* 18 | NP_067027.1 |
HNRNPDL - heterogeneous nuclear ribonucleoprotein D like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001207000.1 | 402 | Intron | NP_001193929.1 | |||
NM_031372.3 | 402 | Intron | NP_112740.1 |