Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601158 | ||||||||||||||||||||
Literature Links: |
MAPK8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAPK8 - mitogen-activated protein kinase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278547.1 | Intron | NP_001265476.1 | ||||
NM_001278548.1 | Intron | NP_001265477.1 | ||||
NM_001323302.1 | Intron | NP_001310231.1 | ||||
NM_001323320.1 | Intron | NP_001310249.1 | ||||
NM_001323321.1 | Intron | NP_001310250.1 | ||||
NM_001323322.1 | Intron | NP_001310251.1 | ||||
NM_001323323.1 | Intron | NP_001310252.1 | ||||
NM_001323324.1 | Intron | NP_001310253.1 | ||||
NM_001323325.1 | Intron | NP_001310254.1 | ||||
NM_001323326.1 | Intron | NP_001310255.1 | ||||
NM_001323327.1 | Intron | NP_001310256.1 | ||||
NM_001323328.1 | Intron | NP_001310257.1 | ||||
NM_001323329.1 | Intron | NP_001310258.1 | ||||
NM_001323330.1 | Intron | NP_001310259.1 | ||||
NM_001323331.1 | Intron | NP_001310260.1 | ||||
NM_139046.3 | Intron | NP_620634.1 | ||||
NM_139049.3 | Intron | NP_620637.1 |