Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTTCGGTATGGTCATGAACTTTA[C/T]TGTTCTCAACTGCATTAAGGGATTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 133510 MIM: 609487 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERCC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ERCC3 - ERCC excision repair 3, TFIIH core complex helicase subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAP3K2 - mitogen-activated protein kinase kinase kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006609.4 | 8488 | UTR 3 | NP_006600.3 |