Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605897 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
UGGT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
UGGT1 - UDP-glucose glycoprotein glucosyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020120.3 | Intron | NP_064505.1 | ||||
XM_006712634.3 | Intron | XP_006712697.1 | ||||
XM_006712635.3 | Intron | XP_006712698.1 | ||||
XM_006712636.3 | Intron | XP_006712699.1 | ||||
XM_017004507.1 | Intron | XP_016859996.1 | ||||
XM_017004508.1 | Intron | XP_016859997.1 | ||||
XM_017004509.1 | Intron | XP_016859998.1 | ||||
XM_017004510.1 | Intron | XP_016859999.1 |
Set Membership: |
HapMap |