Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACAGGCTCCCGCTGTAACTCGCTC[C/T]CGCTGTAACAGGGGCAATCTGGATA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610457 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMPD4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SMPD4 - sphingomyelin phosphodiesterase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171083.2 | Intron | NP_001164554.1 | ||||
NM_017751.4 | Intron | NP_060221.2 | ||||
NM_017951.4 | Intron | NP_060421.2 | ||||
XM_011511443.2 | Intron | XP_011509745.1 | ||||
XM_011511444.2 | Intron | XP_011509746.1 | ||||
XM_011511445.2 | Intron | XP_011509747.1 | ||||
XM_017004448.1 | Intron | XP_016859937.1 | ||||
XM_017004449.1 | Intron | XP_016859938.1 |
Set Membership: |
HapMap |