Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTCCAGTTTTGCACATTTATTGA[A/G]AAGAGGGAATACTATTATAGCATTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CCDC93 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC93 - coiled-coil domain containing 93 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019044.4 | 5640 | UTR 3 | NP_061917.3 | |||
XM_006712600.2 | 5640 | Intron | XP_006712663.1 | |||
XM_011511359.1 | 5640 | UTR 3 | XP_011509661.1 | |||
XM_011511361.1 | 5640 | UTR 3 | XP_011509663.1 | |||
XM_011511362.1 | 5640 | UTR 3 | XP_011509664.1 | |||
XM_011511363.2 | 5640 | Intron | XP_011509665.1 |
LOC101929908 - uncharacterized LOC101929908 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |