Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGGCTTCGGTGACCAGGGCCGTG[C/T]TTGGAGAGCTGCCCTCGGGAGGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 137030 | ||||||||||||||||||||
Literature Links: |
GALM PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GALM - galactose mutarotase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138801.2 | 277 | Missense Mutation | CTT,TTT | L,F 9 | NP_620156.1 | |
XM_011532540.1 | 277 | Missense Mutation | CTT,TTT | L,F 9 | XP_011530842.1 |