Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGAGGTGGCCACAGGACCTTCCA[C/T]GGTCCTTCCCAGGGCACAGTTTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604402 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ST3GAL5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ST3GAL5 - ST3 beta-galactoside alpha-2,3-sialyltransferase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042437.1 | Intron | NP_001035902.1 | ||||
NM_003896.3 | Intron | NP_003887.3 | ||||
XM_005264630.4 | Intron | XP_005264687.1 | ||||
XM_011533143.2 | Intron | XP_011531445.1 | ||||
XM_017005199.1 | Intron | XP_016860688.1 | ||||
XM_017005200.1 | Intron | XP_016860689.1 | ||||
XM_017005201.1 | Intron | XP_016860690.1 | ||||
XM_017005202.1 | Intron | XP_016860691.1 | ||||
XM_017005203.1 | Intron | XP_016860692.1 | ||||
XM_017005204.1 | Intron | XP_016860693.1 | ||||
XM_017005205.1 | Intron | XP_016860694.1 | ||||
XM_017005206.1 | Intron | XP_016860695.1 | ||||
XM_017005207.1 | Intron | XP_016860696.1 | ||||
XM_017005208.1 | Intron | XP_016860697.1 | ||||
XM_017005209.1 | Intron | XP_016860698.1 | ||||
XM_017005210.1 | Intron | XP_016860699.1 | ||||
XM_017005211.1 | Intron | XP_016860700.1 | ||||
XM_017005212.1 | Intron | XP_016860701.1 | ||||
XM_017005213.1 | Intron | XP_016860702.1 | ||||
XM_017005214.1 | Intron | XP_016860703.1 | ||||
XM_017005215.1 | Intron | XP_016860704.1 |
Set Membership: |
HapMap |