Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605163 MIM: 607182 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CXCR6 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CXCR6 - C-X-C motif chemokine receptor 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006564.1 | Intron | NP_006555.1 | ||||
XM_005264809.2 | Intron | XP_005264866.1 | ||||
XM_011533290.2 | Intron | XP_011531592.1 | ||||
XM_011533291.2 | Intron | XP_011531593.1 |
FYCO1 - FYVE and coiled-coil domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024513.3 | Intron | NP_078789.2 | ||||
XM_006713333.3 | Intron | XP_006713396.1 | ||||
XM_006713334.3 | Intron | XP_006713397.1 | ||||
XM_011534111.2 | Intron | XP_011532413.1 |
Set Membership: |
HapMap |