Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCCTAATTCTATCTTGACCACCA[A/G]CATCCCATACTGTGAAACAAATGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601177 MIM: 616519 | ||||||||||||||||||||
Literature Links: |
ARF4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARF4 - ADP ribosylation factor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001660.3 | 551 | Missense Mutation | GCT,GTT | A,V 68 | NP_001651.1 |
PDE12 - phosphodiesterase 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322176.1 | 551 | Intron | NP_001309105.1 | |||
NM_001322177.1 | 551 | Intron | NP_001309106.1 | |||
NM_177966.6 | 551 | Intron | NP_808881.3 |