Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGCACATTGTTGAAGCAGAGGCA[A/G]TGAATTACGAAGGCAGTCCAATTAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 164040 | ||||||||||||||||||||
Literature Links: |
MIR3912 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR3912 - microRNA 3912 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NPM1 - nucleophosmin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037738.2 | Intron | NP_001032827.1 | ||||
NM_002520.6 | Intron | NP_002511.1 | ||||
NM_199185.3 | Intron | NP_954654.1 | ||||
XM_005265920.3 | Intron | XP_005265977.1 | ||||
XM_011534564.1 | Intron | XP_011532866.1 |