Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603291 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BNIP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BNIP1 - BCL2 interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001205.2 | 146 | Missense Mutation | CAA,CAC | Q,H 14 | NP_001196.2 | |
NM_013978.2 | 146 | Missense Mutation | CAA,CAC | Q,H 14 | NP_053581.2 | |
NM_013979.2 | 146 | Missense Mutation | CAA,CAC | Q,H 14 | NP_053582.2 | |
NM_013980.2 | 146 | Missense Mutation | CAA,CAC | Q,H 14 | NP_053583.2 | |
XM_011534638.1 | 146 | Missense Mutation | CAA,CAC | Q,H 14 | XP_011532940.1 | |
XM_011534639.1 | 146 | Missense Mutation | CAA,CAC | Q,H 14 | XP_011532941.1 |
CREBRF - CREB3 regulatory factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |