Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGAGAACCTCTGCAACTTCAGCT[C/T]TGAACCATGTTCTTTTGGGTTTTTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601212 MIM: 617007 | ||||||||||||||||||||
Literature Links: |
PTK2B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PTK2B - protein tyrosine kinase 2 beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004103.4 | Intron | NP_004094.3 | ||||
NM_173174.2 | Intron | NP_775266.1 | ||||
NM_173175.2 | Intron | NP_775267.1 | ||||
NM_173176.2 | Intron | NP_775268.1 | ||||
XM_005273447.4 | Intron | XP_005273504.1 | ||||
XM_005273448.4 | Intron | XP_005273505.1 | ||||
XM_011544441.2 | Intron | XP_011542743.1 | ||||
XM_011544442.2 | Intron | XP_011542744.1 | ||||
XM_017013214.1 | Intron | XP_016868703.1 | ||||
XM_017013215.1 | Intron | XP_016868704.1 | ||||
XM_017013216.1 | Intron | XP_016868705.1 |
TRIM35 - tripartite motif containing 35 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |