Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATCCTGTATTTGGTCTGTTAGCA[A/G]TATTTTTCAGGATTATCCACTTGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609098 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXO25 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FBXO25 - F-box protein 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012173.3 | Intron | NP_036305.2 | ||||
NM_183420.1 | Intron | NP_904356.1 | ||||
NM_183421.1 | Intron | NP_904357.1 | ||||
XM_011534748.2 | Intron | XP_011533050.1 | ||||
XM_011534749.1 | Intron | XP_011533051.1 | ||||
XM_017013313.1 | Intron | XP_016868802.1 | ||||
XM_017013314.1 | Intron | XP_016868803.1 |