Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605333 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDS5B PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PDS5B - PDS5 cohesin associated factor B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015032.3 | Intron | NP_055847.1 | ||||
XM_005266298.4 | Intron | XP_005266355.1 | ||||
XM_011534999.2 | Intron | XP_011533301.1 | ||||
XM_011535000.2 | Intron | XP_011533302.1 | ||||
XM_011535002.2 | Intron | XP_011533304.1 | ||||
XM_017020448.1 | Intron | XP_016875937.1 | ||||
XM_017020449.1 | Intron | XP_016875938.1 | ||||
XM_017020450.1 | Intron | XP_016875939.1 | ||||
XM_017020451.1 | Intron | XP_016875940.1 | ||||
XM_017020452.1 | Intron | XP_016875941.1 | ||||
XM_017020453.1 | Intron | XP_016875942.1 |
Set Membership: |
HapMap |