Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCTTGCATATCCCATGGCAAGAG[C/G]AGGAGCGAGAGAGTGGGGAGGTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615894 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC00909 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LINC00909 - long intergenic non-protein coding RNA 909 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF407 - zinc finger protein 407 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146189.1 | 39 | Intron | NP_001139661.1 | |||
NM_001146190.1 | 39 | Intron | NP_001139662.1 | |||
NM_017757.2 | 39 | Intron | NP_060227.2 | |||
XM_005266726.4 | 39 | Intron | XP_005266783.1 | |||
XM_006722500.3 | 39 | Intron | XP_006722563.1 | |||
XM_011526068.2 | 39 | UTR 5 | XP_011524370.1 | |||
XM_011526069.2 | 39 | Intron | XP_011524371.1 | |||
XM_011526070.1 | 39 | Intron | XP_011524372.1 | |||
XM_017025838.1 | 39 | Intron | XP_016881327.1 |
Set Membership: |
HapMap |