Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGTATTGCAAAGCTTCAGTTCCT[A/C]CTACCGATAAAGTGTGTCTGTGGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300133 | ||||||||||||||||||||
Literature Links: |
VBP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
VBP1 - VHL binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303543.1 | 503 | Missense Mutation | ACT,CCT | T,P 155 | NP_001290472.1 | |
NM_001303544.1 | 503 | Missense Mutation | ACT,CCT | T,P 114 | NP_001290473.1 | |
NM_001303545.1 | 503 | Missense Mutation | ACT,CCT | T,P 82 | NP_001290474.1 | |
NM_003372.6 | 503 | Missense Mutation | ACT,CCT | T,P 119 | NP_003363.1 |