Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAATACGTTCTCATTTCAATCTGG[G/T]TCTTATTAAATATAATTTATGTGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602545 | ||||||||||||||||||||
Literature Links: |
PTPRK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PTPRK - protein tyrosine phosphatase, receptor type K | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135648.2 | 4623 | UTR 3 | NP_001129120.1 | |||
NM_001291981.1 | 4623 | UTR 3 | NP_001278910.1 | |||
NM_001291982.1 | 4623 | Intron | NP_001278911.1 | |||
NM_001291983.1 | 4623 | Intron | NP_001278912.1 | |||
NM_001291984.1 | 4623 | UTR 3 | NP_001278913.1 | |||
NM_002844.3 | 4623 | UTR 3 | NP_002835.2 | |||
XM_006715537.2 | 4623 | UTR 3 | XP_006715600.1 | |||
XM_011536014.2 | 4623 | UTR 3 | XP_011534316.1 | |||
XM_011536015.2 | 4623 | UTR 3 | XP_011534317.1 | |||
XM_011536016.2 | 4623 | UTR 3 | XP_011534318.1 | |||
XM_011536017.2 | 4623 | UTR 3 | XP_011534319.1 | |||
XM_011536018.2 | 4623 | UTR 3 | XP_011534320.1 | |||
XM_011536020.2 | 4623 | Intron | XP_011534322.1 | |||
XM_011536021.2 | 4623 | Intron | XP_011534323.1 | |||
XM_017011145.1 | 4623 | UTR 3 | XP_016866634.1 | |||
XM_017011146.1 | 4623 | UTR 3 | XP_016866635.1 | |||
XM_017011147.1 | 4623 | UTR 3 | XP_016866636.1 | |||
XM_017011148.1 | 4623 | UTR 3 | XP_016866637.1 | |||
XM_017011149.1 | 4623 | UTR 3 | XP_016866638.1 | |||
XM_017011150.1 | 4623 | UTR 3 | XP_016866639.1 |