Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGACTGCTCTTAAAATTGCAAAG[C/T]ATCTTGCTGGAAATGGACTTTTTTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 191163 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100130476 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC100130476 - uncharacterized LOC100130476 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC107986649 - uncharacterized LOC107986649 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNFAIP3 - TNF alpha induced protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270507.1 | Intron | NP_001257436.1 | ||||
NM_001270508.1 | Intron | NP_001257437.1 | ||||
NM_006290.3 | Intron | NP_006281.1 | ||||
XM_005267119.1 | Intron | XP_005267176.1 | ||||
XM_006715555.1 | Intron | XP_006715618.1 | ||||
XM_011536095.1 | Intron | XP_011534397.1 | ||||
XM_011536096.1 | Intron | XP_011534398.1 |
Set Membership: |
HapMap |