Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGCTTCGCGGTGGACAAGGCCTT[C/T]CTCACCTCCCACAAGGGCATCCTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607888 MIM: 605658 MIM: 160710 | ||||||||||||||||||||
Literature Links: |
CMTM5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CMTM5 - CKLF like MARVEL transmembrane domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037288.1 | 517 | Silent Mutation | TTC,TTT | F,F 29 | NP_001032365.1 | |
NM_001288744.1 | 517 | Silent Mutation | TTC,TTT | F,F 29 | NP_001275673.1 | |
NM_001288745.1 | 517 | Silent Mutation | TTC,TTT | F,F 29 | NP_001275674.1 | |
NM_001288746.1 | 517 | Silent Mutation | TTC,TTT | F,F 29 | NP_001275675.1 | |
NM_138460.2 | 517 | Silent Mutation | TTC,TTT | F,F 29 | NP_612469.1 | |
XM_017020953.1 | 517 | Silent Mutation | TTC,TTT | F,F 29 | XP_016876442.1 | |
XM_017020954.1 | 517 | Silent Mutation | TTC,TTT | F,F 29 | XP_016876443.1 |
IL25 - interleukin 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022789.3 | 517 | Intron | NP_073626.1 | |||
NM_172314.1 | 517 | Intron | NP_758525.1 |
MYH6 - myosin heavy chain 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |