Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAACAAAGGCAAAATGAACAGTGA[G/T]GAAAACCTGGAAACAGGACCGAGTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609411 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COX16 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
COX16 - COX16, cytochrome c oxidase assembly homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYNJ2BP - synaptojanin 2 binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018373.2 | 6422 | UTR 3 | NP_060843.2 |
SYNJ2BP-COX16 - SYNJ2BP-COX16 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202547.2 | 6422 | Intron | NP_001189476.1 | |||
NM_001202548.2 | 6422 | Intron | NP_001189477.1 | |||
NM_001202549.2 | 6422 | Intron | NP_001189478.1 |
Set Membership: |
HapMap |