Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGGCCAACAAGAAAAGTCAGGAC[A/C]AAATGACAGAGGATCTGTCCCTGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613279 | ||||||||||||||||||||
Literature Links: |
ZC3H14 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZC3H14 - zinc finger CCCH-type containing 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160103.1 | 356 | Missense Mutation | AAA,CAA | K,Q 46 | NP_001153575.1 | |
NM_001160104.1 | 356 | Missense Mutation | AAA,CAA | K,Q 46 | NP_001153576.1 | |
NM_001326295.1 | 356 | Missense Mutation | AAA,CAA | K,Q 46 | NP_001313224.1 | |
NM_001326296.1 | 356 | Missense Mutation | AAA,CAA | K,Q 46 | NP_001313225.1 | |
NM_001326297.1 | 356 | Missense Mutation | AAA,CAA | K,Q 12 | NP_001313226.1 | |
NM_001326298.1 | 356 | Missense Mutation | AAA,CAA | K,Q 46 | NP_001313227.1 | |
NM_001326299.1 | 356 | Missense Mutation | AAA,CAA | K,Q 46 | NP_001313228.1 | |
NM_001326300.1 | 356 | Intron | NP_001313229.1 | |||
NM_001326301.1 | 356 | Missense Mutation | AAA,CAA | K,Q 12 | NP_001313230.1 | |
NM_001326302.1 | 356 | Intron | NP_001313231.1 | |||
NM_001326303.1 | 356 | Missense Mutation | AAA,CAA | K,Q 12 | NP_001313232.1 | |
NM_001326304.1 | 356 | Intron | NP_001313233.1 | |||
NM_001326305.1 | 356 | Intron | NP_001313234.1 | |||
NM_001326306.1 | 356 | Missense Mutation | AAA,CAA | K,Q 46 | NP_001313235.1 | |
NM_001326307.1 | 356 | Missense Mutation | AAA,CAA | K,Q 46 | NP_001313236.1 | |
NM_001326308.1 | 356 | Intron | NP_001313237.1 | |||
NM_001326309.1 | 356 | Intron | NP_001313238.1 | |||
NM_001326310.1 | 356 | Missense Mutation | AAA,CAA | K,Q 46 | NP_001313239.1 | |
NM_001326311.1 | 356 | Intron | NP_001313240.1 | |||
NM_001326312.1 | 356 | Missense Mutation | AAA,CAA | K,Q 46 | NP_001313241.1 | |
NM_001326313.1 | 356 | Missense Mutation | AAA,CAA | K,Q 46 | NP_001313242.1 | |
NM_001326314.1 | 356 | Intron | NP_001313243.1 | |||
NM_001326315.1 | 356 | Missense Mutation | AAA,CAA | K,Q 12 | NP_001313244.1 | |
NM_001326316.1 | 356 | Missense Mutation | AAA,CAA | K,Q 12 | NP_001313245.1 | |
NM_024824.4 | 356 | Missense Mutation | AAA,CAA | K,Q 46 | NP_079100.2 | |
NM_207660.3 | 356 | Missense Mutation | AAA,CAA | K,Q 46 | NP_997543.1 | |
NM_207661.2 | 356 | Missense Mutation | AAA,CAA | K,Q 12 | NP_997544.1 | |
NM_207662.3 | 356 | Intron | NP_997545.2 | |||
XM_005268067.4 | 356 | Missense Mutation | AAA,CAA | K,Q 46 | XP_005268124.1 | |
XM_005268068.4 | 356 | Missense Mutation | AAA,CAA | K,Q 46 | XP_005268125.1 | |
XM_005268069.4 | 356 | Missense Mutation | AAA,CAA | K,Q 46 | XP_005268126.1 | |
XM_005268070.4 | 356 | Missense Mutation | AAA,CAA | K,Q 46 | XP_005268127.1 | |
XM_005268071.4 | 356 | Missense Mutation | AAA,CAA | K,Q 46 | XP_005268128.1 | |
XM_005268073.3 | 356 | Intron | XP_005268130.1 | |||
XM_006720257.2 | 356 | Intron | XP_006720320.1 | |||
XM_011537160.2 | 356 | Missense Mutation | AAA,CAA | K,Q 12 | XP_011535462.1 | |
XM_011537161.2 | 356 | Missense Mutation | AAA,CAA | K,Q 46 | XP_011535463.1 | |
XM_011537162.2 | 356 | Missense Mutation | AAA,CAA | K,Q 46 | XP_011535464.1 | |
XM_017021648.1 | 356 | Missense Mutation | AAA,CAA | K,Q 46 | XP_016877137.1 | |
XM_017021649.1 | 356 | Missense Mutation | AAA,CAA | K,Q 46 | XP_016877138.1 | |
XM_017021650.1 | 356 | Missense Mutation | AAA,CAA | K,Q 46 | XP_016877139.1 |