Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCGCTGCTGCCCATCTTCTTCGG[C/T]GCCCTGCGCTCCGTACGCTGCGCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607106 | ||||||||||||||||||||
Literature Links: |
HM13 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HM13 - histocompatibility minor 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030789.3 | 292 | Silent Mutation | GGC,GGT | G,G 50 | NP_110416.1 | |
NM_178580.2 | 292 | Silent Mutation | GGC,GGT | G,G 50 | NP_848695.1 | |
NM_178581.2 | 292 | Silent Mutation | GGC,GGT | G,G 50 | NP_848696.1 | |
NM_178582.2 | 292 | Silent Mutation | GGC,GGT | G,G 50 | NP_848697.1 |